Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GMHGGHTFK PLAEIYEQHV TKVNEEVAKL RRRLMELISL VQEVERNVEA VRNAKDERVR EIRNAVEMMI ARLDTQLKNK LITLMGQKTS LTQETELLES LLQEVEHQLR SCSKSELISK SSEILMMFQQ VHRKPMASFV TTPVPPDFTS ELVPSYDSAT FVLENFSTLR QRADPVYSPP LQVSGLCWRL KVYPDGNGVV RGYYLSVFLE LSAGLPETSK YEYRVEMVHQ SCNDPTKNII REFASDFEVG ECWGYNRFFR LDLLANEGYL NPQNDTVILR FQVRSPTFFQ KSRDQHWYIT QLEAAQTSYI QQINNLKERL TIELSRTQKS (122-450 aa encoded by BC036012)
Phosphorylates phosphatidylinositol (PI) in the first committed step in the production of the second messenger inositol-1, 4, 5, -trisphosphate (PIP). PI4KB may regulate Golgi disintegration/reorganization during mitosis, possibly via its phosphorylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM37; Mulibrey nanism protein; RING-B-box-coiled-coil protein; RING-type E3 ubiquitin transferase TRIM37; TRAF encompassing factor 3; Tripartite motif-containing protein 37
Gene Aliases: 1110032A10Rik; 2810004E07Rik; AI848587; AU043018; KIAA0898; mKIAA0898; MUL; POB1; TEF3; TRIM37
UniProt ID: (Human) O94972, (Mouse) Q6PCX9
Entrez Gene ID: (Human) 4591, (Mouse) 68729
Molecular Function:
ubiquitin-protein ligase
protein modifying enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support