Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MSASLNYKSF SKEQQTMDNL EKQLICPICL EMFTKPVVIL PCQHNLCRKC ASDIFQASNP YLPTRGGTTM ASGGRFRCPS CRHEVVLDRH GVYGLQRNLL VENIIDIYKQ ESTRPEKKSD QPMCEEHEEE RINIYCLNCE VPTCSLCKVF GAHKDCQVAP LTHVFQRQKS ELSDGIAILV GSNDRVQGVI SQLEDTCKTI EECCRKQKQE LCEKFDYLYG ILEERKNEMT QVITRTQEEK LEHVRALIKK YSDHLENVSK LVESGIQFMD EPEMAVFLQN AKTLLKKISE ASKAFQMEKI EHGYENMNHF TVNLNREEKI IREIDFYRED EDEEEEEGGE GEKEGEGEVG GEAVEVEEVE NVQTEFPGED ENPEKASELS QVELQAAPGA LPVSSPEPPP ALPPAADAPV TQIGFEAPPL QGQAAAPASG SGADSEPARH IFSFSWLNSL NE
The protein encoded by this gene contains a RING zinc finger, a motif known to be involved in protein-protein interactions. This protein associates transiently with microtubules, myosin, and titin during muscle sarcomere assembly. It may act as a transient adaptor and plays a regulatory role in the assembly of sarcomeres. Four alternatively spliced transcript variants encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MuRF-2; muscle specific ring finger 2; Muscle-specific RING finger protein 2; RING finger protein 29; tripartite motif-containing 55; Tripartite motif-containing protein 55
Gene Aliases: MURF-2; MURF2; RNF29; TRIM55
UniProt ID: (Human) Q9BYV6
Entrez Gene ID: (Human) 84675
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support