Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLRNILDRV EQRELKKLEQ EEKKGLRIIE EAENDLVHQT QSLRELISDL ERRCQGSTME LLQDVSDVTE RSEFWTLRKP EALPTKLRSM FRAPDLKRML RVCRELTDVQ SYWVDVTLNP HTANLNLVLA KNRRQVRFVG AKVSGPSCLE KHYDCSVLGS QHFSSGKHYW EVDVAKKTAW ILGVCSNSLG PTFSFNHFAQ NHSAYSRYQP QSGYWVIGLQ HNHEYRAYED SSPSLLLSMT VPPRRVGVFL DYEAGTVSFY NVTNHGFPIY TFSKYYFPTT LCPYFNPCNC VIPMTLRRPS S (96-395 aa encoded by BC065575)
TRIM6 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified.The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to the nucleus, but its specific function has not been identified. This gene is mapped to chromosome 11p15, where it resides within a TRIM gene cluster. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. A read-through transcript transcribed from this gene and TRIM34 has been observed.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 89; RING-type E3 ubiquitin transferase TRIM6; tripartite motif-containing 6; Tripartite motif-containing protein 6
Gene Aliases: RNF89; TRIM6
UniProt ID: (Human) Q9C030
Entrez Gene ID: (Human) 117854
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support