Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thyroid hormone receptors (TRs) are transcription factors that regulate the expression of specific genes in a hormone-dependent manner. TRIP13 (thyroid hormone receptor interactor 13), also called 16E1BP, is a transcription factor that interacts with the ligand binding domain of the thyroid receptor (TR) as well as a variety of target genes including human papilloma virus type 16 (HPV16) E1. Unlike most TRIP proteins which function only in the presence of hormones, TRIP13 does not require the presence of thyroid hormone to interact with TR. The association of TRIP13 with (HPV16) E1 suggests that TRIP13 may have tumor suppressor gene function. TRIP13 is a 432 amino acid protein with 2 different isoforms produced by alternative splicing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 16E1-BP; HPV16 E1 protein binding protein; HPV16 E1 protein-binding protein; Human papillomavirus type 16 E1 protein-binding protein; Pachytene checkpoint protein 2 homolog; Thyroid hormone receptor interactor 13; thyroid receptor interacting protein 13; Thyroid receptor-interacting protein 13; TR-interacting protein 13; TRIP-13
Gene Aliases: 16E1BP; 2410002G23Rik; D13Ertd328e; PCH2; TRIP13
UniProt ID: (Human) Q15645, (Rat) Q5XHZ9, (Mouse) Q3UA06
Entrez Gene ID: (Human) 9319, (Rat) 292206, (Mouse) 69716
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support