Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TFNKTLTQE EKIKRIDALE FLPFEGKVNL KKPQHVFSVL EDYGLDPNCI PENPHNIYFG RWIADGQREL IESYSVKKRH FIGNTSMDAG LSFIMANHGK VKENDIVFDP FVGTGGLQIA CAHFGAYVYG TDIDYNTVHG LGKATRKNQK WRGPDENIRA NLRQYGLEKY YLDVLVSDAS KPSWRKGTYF DAIITDPPYG IRESTRRTGS QKEIPKGIEK WEKCPESHVP VSLSYHLSDM FLDLLNFAAE TLVLGGRLVY WLPVYTPEYT EEMVPWHPCL ELVSNCEQKL SSHTSRRLIT MEKVKKFENR DQYSHLLSDH FLPYQGHNSF REKYFSGVTK RIAKEEKSTQ E (114-463 aa encoded by BC056878)
TRMT11 belongs to the methyltransferase superfamily. It is a catalytic subunit of an S-adenosyl-L-methionine-dependent tRNA methyltransferase complex that mediates the methylation of the guanosine nucleotide at position 10 (m2G10) in tRNAs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: putative RNA methylase MDS024; tRNA (guanine(10)-N2)-methyltransferase homolog; tRNA guanosine-2'-O-methyltransferase TRM11 homolog; tRNA methyltransferase 11 homolog
Gene Aliases: 2410075D05Rik; 3110045I18Rik; AW213713; C6orf75; MDS024; TRM11; TRMT11; TRMT11-1
UniProt ID: (Human) Q7Z4G4, (Rat) Q7TNK6
Entrez Gene ID: (Human) 60487, (Mouse) 73681, (Rat) 378794
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support