Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: QLNVSPQIS KYNLELTYEH FKSEEILRAV LPEGQDVTSG FSRIGHIAHL NLRDHQLPFK HLIGQVMIDK NPGITSAVNK INNIDNMYRN FQMEVLSGEQ NMMTKVRENN YTYEFDFSKV YWNPRLSTEH SRITELLKPG DVLFDVFAGV GPFAIPVAKK NCTVFANDLN PESHKWLLYN CKLNKVDQKV KVFNLDGKDF LQGPVKEELM QLLGLSKERK PSVHVVMNLP AKAIEFLSAF KWLLDGQPCS SEFLPIVHCY SFSKDANPAE DVRQRAGAVL GISLEACSSV HLVRNVAPNK EMLCITFQIP ASVLYKNQTR NPENHEDPPL KRQRTAEAFS DEKTQIVSNT (161-509 aa encoded by BC108284)
tRNAs contain as many as 13 or 14 nucleotides that are modified posttranscriptionally by enzymes that are highly specific for particular nucleotides in the tRNA structure. TRMT5 methylates the N1 position of guanosine-37 in selected tRNAs using S-adenosyl methionine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M1G-methyltransferase; TRM5 tRNA methyltransferase 5 homolog; TRMT5; tRNA (guanine(37)-N1)-methyltransferase; tRNA (guanine-N(1)-)-methyltransferase; tRNA [GM37] methyltransferase; tRNA methyltransferase 5 homolog; tRNA-N1G37 methyltransferase
Gene Aliases: 2610027O18Rik; COXPD26; KIAA1393; mKIAA1393; RGD1306567; TRM5; TRMT5
UniProt ID: (Human) Q32P41, (Mouse) Q9D0C4
Entrez Gene ID: (Human) 57570, (Mouse) 76357, (Rat) 362754
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support