Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to 56 kDa TRMT6 protein.
Immunogen sequence: MEGSGEQPG PQPQHPGDHR IRDGDFVVLK REDVFKAVQV QRRKKVTFEK QWFYLDNVIG HSYGTAFEVT SGGSLQPKKK REEPTAETKE AGTDNRNIVD DGKSQKLTQD DIKALKDKGI KGEEIVQQLI ENSTTFRDKT EFAQDKYIKK KKKKYEAIIT VVKPSTRILS IMYYAREPGK INHMRYDTLA QMLTLGNIRA GNKMIVMETC AGLVLGAMME RMGGFGSIIQ LYPGGGPVRA ATACFGFPKS FLSGLYEFPL NKVDSLLHGT FSAKMLSSEP KDSALVEESN GTLEEKQASE QENEDSMAEA PESNHPEDQE TMETISQDPE HKGPKERGSK KDYIQEKQRR QEEQRKRHLE AAALLSERNA DGLIVASRFH PTPLLLSLLD FVAPSRPFVV YCQYKEPLLE CYTKLRERGG VINLRLSETW LRNYQVLPDR SHPKLLMSGG GGYLLSGFTV AMDNLKADTS LKSNASTLES HETEEPAAKK RKCPESDS (1-497 aa encoded by BC001262)
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC5029; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6
Gene Aliases: 3300001M20Rik; AU016412; AW492787; CGI-09; GCD10; Gcd10p; KIAA1153; mKIAA1153; RGD1308877; TRM6; TRMT6
UniProt ID: (Human) Q9UJA5, (Mouse) Q8CE96
Entrez Gene ID: (Human) 51605, (Rat) 311441, (Mouse) 66926
Molecular Function:
translation factor
translation initiation factor
translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support