Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: MEGSGEQPGP QPQHPGDHRI RDGDFVVLKR EDVFKAVQVQ RRKKVTFEKQ WFYLDNVIGH SYGTAFEVTS GGSLQPKKKR EEPTAETKEA GTDNRNIVDD GKSQKLTQDD IKALKDKGIK GEEIVQQLIE NSTTFRDKTE FAQDKYIKKK KKKYEAIITV VKPSTRILSI MYYAREPGKI NHMRYDTLAQ MLTLGNIRAG; Positive Samples: U-87MG, HepG2; Cellular Location: Nucleus
Substrate-binding subunit of tRNA (adenine-N(1)-)-methyltransferase, which catalyzes the formation of N(1)-methyladenine at position 58 (m1A58) in initiator methionyl-tRNA.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC5029; mRNA methyladenosine-N(1)-methyltransferase non-catalytic subunit TRM6; tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6; tRNA methyltransferase 6 homolog; tRNA(m1A58)-methyltransferase subunit TRM6; tRNA(m1A58)MTase subunit TRM6
Gene Aliases: CGI-09; GCD10; Gcd10p; KIAA1153; TRM6; TRMT6
UniProt ID: (Human) Q9UJA5
Entrez Gene ID: (Human) 51605
Molecular Function: translation factor translation initiation factor translational protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support