Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAASLWMGD LEPYMDENFI SRAFATMGET VMSVKIIRNR LTGIPAGYCF VEFADLATAE KCLHKINGKP LPGATPAKRF KLNYATYGKQ PDNSPEYSLF VGDLTPDVDD GMLYEFFVKV YPSCRGGKVV LDQTGVSKGY GFVKFTDELE QKRALTECQG AVGLGSKPVR LSVAIPKASR VKPVEYSQMY SYSYNQYYQQ YQNYYAQWGY DQNTGSYSYS YPQYGYTQST MQTYEEVGDD ALEDPMPQLD VTEANKEFME QSEELYDALM DCHWQPLDTV SSEIPAMM (1-287 aa encoded by BC000680)
TRSPAP1 is involved in the early steps of selenocysteine biosynthesis and tRNA(Sec) charging to the later steps resulting in the cotranslational incorporation of selenocysteine into selenoproteins. It stabilizes the SECISBP2, EEFSEC and tRNA(Sec) complex. It may be involved in the methylation of tRNA(Sec) and enhances efficiency of selenoproteins synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ20503; RP4-669K10.4; SECp43; tRNA selenocysteine 1-associated protein 1; tRNA selenocysteine associated protein; tRNA selenocysteine associated protein (SECP43); tRNA selenocysteine associated protein 1; tRNA selenocysteine-associated protein 1
Gene Aliases: 1110007F05Rik; AA387999; PRO1902; SECP43; TRNAU1AP; TRSPAP1; WI1-2643G14.2
UniProt ID: (Human) Q9NX07, (Mouse) Q80VC6, (Rat) Q9QZI7
Entrez Gene ID: (Human) 54952, (Mouse) 71787, (Rat) 65241
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support