Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MEESVNQMQP LNEKQIANSQ DGYVWQVTDM NRLHRFLCFG SEGGTYYIKE QKLGLENAEA LIRLIEDGRG CEVIQEIKSF SQEGRTTKQE PMLFALAICS
TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; gastric cancer multi-drug resistance protein; RNA-binding protein RO60; Ro 60 kDa autoantigen; Ro60 autoantigen; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); SS-A; TROVE domain family member 2; TROVE domain family, member 2
Gene Aliases: RO60; RORNP; SSA2; TROVE2
UniProt ID: (Human) P10155
Entrez Gene ID: (Human) 6738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support