Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MEESVNQMQL LNEKQIANSQ DGYVWQVTDM NRLHRFLCFG SEGGTYYIKE QKLGLENAEA LIRLIEDGRG CEVIQEIKSF SQEGRTTKQE PMLFALAICS QCSDISTKQA AFKAVSEVCR IPTHLFTFIQ FKKDLKESMK CGMWGRALRK AIADWYNEKG GMALALAVTK YKQRNGWSHK DLLRLSHLKP SSEGLAIVTK YITKGWKEVH ELYKEKALSV ETEKLLKYLE AVEKVKRTRD ELEVIHLIEE HRLVREHLLT NHLKSKEVWK ALLQETPLTA LLRNLGKMTA NSVLEPGNSE VSLVCEKLCN EKLLKKARIH PFHILIALET YKTGHGLRGK LKWRPDEEIL KALDAAFYKT FKTVEPTGKR FLLAVDVSAS MNQRVLGSIL NASTVAAAMC MVVTRTEKDS YVVAFSDEMV PCPVTTDMTL QQVLMAMSQI PAGGTDCSLP MIWAQKTNTP ADVFIVFTDN ETFAGGVHPA IALREYRKKM DIPAKLIVCG MTSNGFTIAD PDDRGMLDMC GFDTGALDVI RNFTLDMI
TROVE2 belongs to the Ro 60 kDa family. It is RNA-binding protein that binds to several small cytoplasmic RNA molecules known as Y RNAs. It may stabilize these RNAs from degradation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 60 kDa ribonucleoprotein Ro; 60 kDa Ro protein; 60 kDa SS-A/Ro ribonucleoprotein; gastric cancer multi-drug resistance protein; RNA-binding protein RO60; Ro 60 kDa autoantigen; Ro60 autoantigen; RP11-101E13.3; Sjoegren syndrome antigen A2; Sjoegren syndrome type A antigen; Sjogren syndrome antigen A2 (60kD, ribonucleoprotein autoantigen SS-A/Ro); SS-A; TROVE domain family member 2; TROVE domain family, member 2
Gene Aliases: RO60; RORNP; SSA2; TROVE2
UniProt ID: (Human) P10155
Entrez Gene ID: (Human) 6738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support