Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: TRPA1.
TRPA1 is a TRP-related channel that responds to cold temperatures and pungent compounds and plays a role in both nociceptor and hair cell transduction. TRPA1 is a transformation -associated gene product in lung epithelia, whereas its protein distribution is primarily restricted to sensory neurons. Blocking TRPA1 may be a therapeutic target for treating cold hyperalgesia caused by inflammation and nerve damage. It is now known that TRPA1 protein is also widely expressed outside of the CNS and is dys-regulated during oncogenic transformation. The structure of TRPA1 is highly related to both the protein ankyrin and transmembrane proteins. The specific function of TRPA1 has not yet been determined; however, studies indicate the function may involve a role in signal transduction and growth control.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ankyrin-like with transmembrane domains protein 1; CG5751-PG; CG5751-PH; CG5751-PI; CG5751-PJ; CG5751-PK; dANKTM1; dANKTM1; TRP ion channel family member; dTRPA1; nompC; transient receptor potential A 1; transient receptor potential A1; transient receptor potential cation channel; transient receptor potential cation channel A1 ortholog; Transient receptor potential cation channel subfamily A member 1; transient-receptor-potential A1; TRPA-1; TrPA1; TrpA1-PG; TrpA1-PH; TrpA1-PI; TrpA1-PJ; TrpA1-PK
Gene Aliases: ANKTM1; CG5751; CG5761; CT18073; dANKTM1; Dmel\CG5751; Dmel_CG5751; DmTRPA1; dTRPA; dTRPA1; Trp1A; TRPA; TRPA1
UniProt ID: (Fruit fly) Q7Z020
Entrez Gene ID: (Fruit fly) 39015
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support