Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MQDVQGPRP GSPGDAEDRR ELGLHRGEVN FGGSGKKRGK FVRVPSGVAP SVLFDLLLAE WHLPAPNLVV SLVGEEQPFA MKSWLRDVLR KGLVKAAQST GAWILTSALR VGLARHVGQA VRDHSLASTS TKVRVVAVGM ASLGRVLHRR ILEEAQEDFP VHYPEDDGGS QGPLCSLDSN LSHFILVEPG PPGKGDGLTE LRLRLEKHIS EQRAGYGGTG SIEIPVLCLL VNGDPNTLER ISRAVEQAAP WLILAGSGGI ADVLAALVNQ PHLLVPKVAE KQFKEKFPSK HFSWEDIVRW TKLLQNITSH QHLLTVYDFE QEGSEELDTV ILKALVKACK SHSQEPQDYL D (1-350 aa encoded by BC093787)
This gene encodes a member of the transient receptor potential protein family, which is a diverse group of proteins with structural features typical of ion channels. This protein plays an important role in taste transduction, and has characteristics of a calcium-activated, non-selective cation channel that carries Na+, K+, and Cs+ ions equally well, but not Caions. It is activated by lower concentrations of intracellular Ca, and inhibited by higher concentrations. It is also a highly temperature-sensitive, heat activated channel showing a steep increase of inward currents at temperatures between 15 and 35 degrees Celsius. This gene is located within the Beckwith-Wiedemann syndrome critical region-1 on chromosome 11p15. 5, and has been shown to be imprinted, with exclusive expression from the paternal allele.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Long transient receptor potential channel 5; long transient receptor potential-related channel 5; LTrpC-5; MLSN1 and TRP-related; MLSN1- and TRP-related gene 1 protein; Transient receptor potential cation channel subfamily M member 5; transient receptor potential cation channel, subfamily M, member 5; TRPM5
Gene Aliases: 9430099A16Rik; LTrpC-5; LTRPC5; MTR1; TRPM5
UniProt ID: (Human) Q9NZQ8, (Mouse) Q9JJH7
Entrez Gene ID: (Human) 29850, (Mouse) 56843, (Rat) 365391
Molecular Function:
ion channel
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support