Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.
Specificity of this antibody: TRPN1.
TRPN1 is a candidate mechanotransduction channel in Drosophila and Caenorhabditis elegans, also present in hair cells of lower vertebrates. At its N-terminal cytoplasmic tail it contains 28 ankyrin repeats. (PMID: 23159937)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: no mechanoreceptor potential C
Gene Aliases: nompC
Entrez Gene ID: (Zebrafish) 368273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support