Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
1 µg/mL of MA5-45457 was sufficient for detection of TrpV3 in 10 µg of COS-1 cell lysate transiently transfected with TrpV3 by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody|Detects approximately 70kDa.
TRPV3 belongs to a family of nonselective cation channels that function in a variety of processes, including temperature sensation and vasoregulation. The thermosensitive members of this family are expressed in subsets of sensory neurons that terminate in the skin, and are activated at distinct physiological temperatures. This channel is activated at temperatures between 22 and 40 degrees C. This gene lies in close proximity to another family member (TRPV1) gene on chromosome 17, and the two encoded proteins are thought to associate with each other to form heteromeric channels.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: heat sensitive channel TRPV3; MGC124324; MGC124325; non-hair; Transient receptor potential cation channel subfamily V member 3; transient receptor potential cation channel, subfamily V, member 3; TRP ion channel TRPV3; TrpV3; Vanilloid receptor-like 3; vanilloid receptor-related osmotically activated channel protein; VRL 3; VRL-3
Gene Aliases: 1110036I10Rik; AI644701; FNEPPK2; Nh; OLMS; TRPV3; VRL3
UniProt ID: (Human) Q8NET8, (Mouse) Q8K424
Entrez Gene ID: (Human) 162514, (Mouse) 246788, (Rat) 497948
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support