Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EEEGLVERRE EAQRAQQAVP GPGPMTPESA LEELLAVQVE LEPVNAQARK AF
Highest antigen sequence identity to the following orthologs: Mouse - 37%, Rat - 38%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Testis-specific Y-encoded protein 4
Gene Aliases: TSPY10; TSPY4; TSPY8
UniProt ID: (Human) P0CV99
Entrez Gene ID: (Human) 728395
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support