Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MDDAAVLKRR GYLLGINLGE GSYAKVKSAY SERLKFNVAI KIIDRKKAPA DFLEKFLPRE IEILAMLNHC SIIKTYEIFE TSHGKVYIVM ELAVQGDLLE LIKTRGALHE DEARKKFHQL SLAIKYCHDL DVVHRDLKCD NLLLDKDFNI KLSDFSFSKR CLRDDSGRMA LSKTFCGSPA YAAPEVLQGI PYQPKVYDIW SLGVILYIMV CGSMPYDDSN IKKMLRIQKE HRVNFPRSKH LTGECKDLIY HMLQPDVNRR LHIDEILSHC WMQPKARGSP SVAINKEGES SRGTEPLWTP EPGSDKKSAT KLEPEGEAQP QAQPETKPEG TAMQMSRQSE ILGFPSKPST METEEGPPQQ PPETRAQ
STK22D (TSSK1) is a serine/threonine kinase essential to the function and production of sperm (spermiogenesis). TSSK1 belongs to a family of serine/threonine kinases highly expressed in testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: serine/threonine kinase 22D (spermiogenesis associated); serine/threonine kinase FKSG81; Serine/threonine-protein kinase 22A; spermiogenesis associated 4; testis tissue sperm-binding protein Li 55e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1B; Testis-specific serine/threonine-protein kinase 1; TSK-1; TSSK-1
Gene Aliases: FKSG81; SPOGA1; SPOGA4; STK22A; STK22D; TSK1; TSSK1; TSSK1B
UniProt ID: (Human) Q9BXA7
Entrez Gene ID: (Human) 83942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support