Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LSHCWMQPKA RGSPSVAINK EGESSRGTEP LWTPEPGSDK KSATKLEPEG EAQPQAQPET KPEGTAMQMS RQSEILGFPS KPSTMETEEG PPQQPPETRA Q
STK22D (TSSK1) is a serine/threonine kinase essential to the function and production of sperm (spermiogenesis). TSSK1 belongs to a family of serine/threonine kinases highly expressed in testis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: serine/threonine kinase 22A (spermiogenesis associated); serine/threonine kinase 22D (spermiogenesis associated); serine/threonine kinase FKSG81; Serine/threonine-protein kinase 22A; spermiogenesis associated 4; testis tissue sperm-binding protein Li 55e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1B; testis-specific serine/threonine kinase; Testis-specific serine/threonine-protein kinase 1; TSK-1; TSSK-1
Gene Aliases: FKSG81; SPOGA1; SPOGA4; STK22A; STK22D; TSK-1; TSK1; Tssk; TSSK1; TSSK1B
UniProt ID: (Human) Q9BXA7, (Mouse) Q61241
Entrez Gene ID: (Human) 83942, (Mouse) 22114
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support