Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: EILSHSWLQP PKPKATSSAS FKREGEGKYR AECKLDTKTD LRPDHRPDHK LGAKTQHRLL VVPENENRME DRLAETSRAK DHHISGAEVG KAST
STK22B (TSSK2) is a serine/threonine kinase essential for the function and production of sperm (spermiogenesis). Deletions in the STK22B coding region correlate with numerous cardiovascular and glandular malformations. TSSK2 gene ontology annotations related to this gene include intracellular signal transduction; peptidyl-serine phosphorylation; protein autophosphorylation; protein phosphorylation; spermatid development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DGS-G; DiGeorge syndrome protein G; serine/threonine kinase 22B (spermiogenesis associated); Serine/threonine-protein kinase 22B; spermiogenesis associated 2; testicular secretory protein Li 61; testis specific serine threonine kinase 2; testis-specific kinase 2; testis-specific serine kinase 2; Testis-specific serine/threonine-protein kinase 2; TSK-2; TSSK-2
Gene Aliases: DGS-G; DGSG; SPOGA2; STK22B; TSK2; TSSK2
UniProt ID: (Human) Q96PF2
Entrez Gene ID: (Human) 23617
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support