Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MVHQVLYRA LVSTKWLAES IRTGKLGPGL RVLDASWYSP GTREARKEYL ERHVPGASFF DIEECRDTAS PYEMMLPSEA GFAEYVGRLG ISNHTHVVVY DGEHLGSFYA PRVWWMFRVF GHRTVSVLNG GFRNWLKEGH PVTSEPSRPE PAVFKATLDR SLLKTYEQVL ENLESKRFQL VDSRSQGRFL GTEPEPDAVG LDSGHIRGAV NMPFMDFLTE DGFEKGPEEL RALFQTKKVD LSQPLIATCR KGVTACHVAL AAYLCGKPDV AVYDGSWSEW FRRAPPESRV SQGKSEKA (1-297 aa encoded by BC010148)
The product of this gene is a mitochondrial matrix enzyme that is encoded by the nucleus. It may play roles in cyanide detoxification, the formation of iron-sulfur proteins, and the modification of sulfur-containing enzymes. The gene product contains two highly conservative domains (rhodanese homology domains), suggesting these domains have a common evolutionary origin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: LL22NC01-146D10.3; MGC19578; Rhodanese; Thiosulfate sulfurtransferase; thiosulfate sulfurtransferase (rhodanese); thiosulfate sulfurtransferase, mitochondrial; Thiosulfate sulphurtransferase (rhodanese)
Gene Aliases: RDS; RHODAN; Rhodanese; TST
UniProt ID: (Human) Q16762, (Rat) P24329, (Mouse) P52196
Entrez Gene ID: (Human) 7263, (Rat) 25274, (Mouse) 22117
Molecular Function:
transferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support