Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: ALAHEPVNEL SALMEDGRCQ VLLAKVYSKM EKLGDAITAL QQARELQARV LKRVQMEQPD AVPAQKHLAA EICAEIAKHS VAQRDYE
Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.
The tetratricopeptide repeat (TPR) motif is a degenerate, 34 amino acid sequence found in many proteins and acts to mediate protein-protein interactions in various pathways. At the sequence level, there can be up to 16 tandem TPR repeats, each of which has a helix-turn-helix shape that stacks on other TPR repeats to achieve ligand binding specificity. TTC21B (tetratricopeptide repeat domain 21B), also known as THM1, is a 1,316 amino acid protein that contains 19 TPR repeats and belongs to the TTC21 family. Localizing to cytoplasm and cytoskeleton, TTC21B exists as two alternatively spliced isoforms and is thought to negatively regulate Shh signal transduction. TTC21B may also be involved in retrograde intraflagellar transport in cilia, and is encoded by a gene that maps to human chromosome 2q24.3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intraflagellar transport 139 homolog; putative protein product of Nbla10696; Tetratricopeptide repeat protein 21B; TPR repeat protein 21B
Gene Aliases: ATD4; IFT139; IFT139B; JBTS11; KIAA1992; Nbla10696; NPHP12; SRTD4; THM1; TTC21B
UniProt ID: (Human) Q7Z4L5
Entrez Gene ID: (Human) 79809
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support