Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide sequence: IQLYDRILQE DPTNTAARKR KIAIRKAQGK NVEAIRELNE YLEQFVGDQE
Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
The exact function of TTC35 remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ER membrane protein complex subunit 2; Tetratricopeptide repeat protein 35; TPR repeat protein 35
Gene Aliases: EMC2; KIAA0103; TTC35
UniProt ID: (Human) Q15006
Entrez Gene ID: (Human) 9694
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support