Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tetratricopeptide repeat domain 7; Tetratricopeptide repeat protein 7A; TPR repeat protein 7A
Gene Aliases: 1110035E02Rik; 1700007L07Rik; fsn; GIDID; hea; KIAA1140; MINAT; TTC7; TTC7A
UniProt ID: (Human) Q9ULT0, (Mouse) Q80XT5
Entrez Gene ID: (Human) 57217, (Mouse) 225049, (Rat) 362696
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support