Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tetratricopeptide repeat domain 7; Tetratricopeptide repeat protein 7A; TPR repeat protein 7A
Gene Aliases: GIDID; KIAA1140; MINAT; TTC7; TTC7A
UniProt ID: (Human) Q9ULT0
Entrez Gene ID: (Human) 57217, (Rat) 362696
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support