Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LQESNLIMA KLNYVEGDYK EALNIYARVG LDDLPLTAVP PYRLRVIAEA YATKGLCLEK LPISSSTSNL HVDREQDVIT CYEKAGDIAL LYLQEIERVI LSNIQNRSPK PGPAPHDQEL GFFLETGLQR AHVLYFKNGN LTRGVGRFRE LLRAVETRTT QNLRMTIARQ LAEILLRGMC EQSYWNPLED PPCQSPLDDP LRKGANTKTY TLTRRARVYS GENI (96-318 aa encoded by BC153122)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tetratricopeptide repeat domain 7 like 1; Tetratricopeptide repeat protein 7-like-1; Tetratricopeptide repeat protein 7B; TPR repeat protein 7-like-1; TPR repeat protein 7B
Gene Aliases: AA408451; c14_5685; TTC7B; TTC7L1
UniProt ID: (Human) Q86TV6, (Mouse) E9Q6P5
Entrez Gene ID: (Human) 145567, (Mouse) 104718
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support