Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
TTF1 is a transcription termination factor that is localized to the nucleolus and plays a critical role in ribosomal gene transcription. The protein mediates the termination of RNA polymerase I transcription by binding to Sal box terminator elements downstream of pre-rRNA coding regions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BCH; BHC; NK-2; NK-2 homolog A; NKX2; NKX2.1; NKX2A; RNA polymerase I termination factor; TEBP; TITF1; Transcription termination factor 1; Transcription termination factor I; transcription termination factor, RNA polymerase I; TTF-1; TTF-I
Gene Aliases: TTF-1; TTF-I; TTF1
UniProt ID: (Human) Q15361
Entrez Gene ID: (Human) 7270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support