Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MYTFVVRDE NSSVYAEVSR LLLATGHWKR LRRDNPRFNL MLGERNRLPF GRLGHEPGLV QLVNYYRGAD KLCRKASLVK LIKTSPELAE SCTWFPESYV IYPTNLKTPV APAQNGIQPP ISNSRTDERE FFLASYNRKK EDGEGNVWIA KSSAGAKGEG ILISSEASEL LDFIDNQGQV HVIQKYLEHP LLLEPGHRKF DIRSWVLVDH QYNIYLYREG VLRTASEPYH VDNFQDKTCH LTNHCIQKEY SKNYGKYEEG NEMFFKEFNQ YLTSALNITL ESSILLQIKH IIRNCLLSVE PAISTKHLPY QSFQLFGFDF MVDEELKVWL IEVNGAPACA QKLYAELCQG IVDIAISSVF PPPDVEQPQT QPAAFIKL (1-377 aa encoded by BC036819)
Catalyzes the post-translational addition of a tyrosine to the C-terminal end of detyrosinated alpha-tubulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2410003M22Rik; TTL; Tubulin--tyrosine ligase; tubulin-tyrosine ligase
Gene Aliases: 2410003M22Rik; 2700049H19Rik; AI848570; TTL
UniProt ID: (Pig) P38160, (Human) Q8NG68, (Mouse) P38585, (Rat) Q9QXJ0
Entrez Gene ID: (Pig) 445530, (Human) 150465, (Mouse) 69737, (Rat) 171572
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support