Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Peptide sequence: QVLQRPPPTL PPSKPKPVQG LCPHGKPRDK GRSCKRSSGH GSGENGSQRP
Sequence homology: Human: 100%; Rabbit: 92%
TTLL11 is a polyglutamase which preferentially modifies alpha-tubulin. It is involved in the side-chain elongation step of the polyglutamylation reaction rather than in the initiation step. TTLL11 is required for CCSAP localization to both spindle and cilia microtubules.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tubulin polyglutamylase TTLL11; tubulin tyrosine ligase like11; tubulin tyrosine ligase-like family member 11; tubulin tyrosine ligase-like family, member 11; Tubulin--tyrosine ligase-like protein 11
Gene Aliases: bA244O19.1; C9orf20; TTLL11
UniProt ID: (Human) Q8NHH1
Entrez Gene ID: (Human) 158135
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support