Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 9
Antibody detects endogenous levels of total TTLL6.
Predicted to enable tubulin binding activity and tubulin-glutamic acid ligase activity. Predicted to be involved in microtubule cytoskeleton organization and protein polyglutamylation. Predicted to act upstream of or within microtubule bundle formation; microtubule severing; and positive regulation of cilium movement. Located in ciliary basal body.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: polyglutamylase; Protein polyglutamylase TTLL6; TTLL6; Tubulin polyglutamylase TTLL6; tubulin tyrosine ligase-like family member 6; tubulin tyrosine ligase-like family, member 6; tubulin tyrosine ligase-like family, member 6, pseudogene 1; Tubulin--tyrosine ligase-like protein 6
Gene Aliases: 4932418K24Rik; D11Moh43e; D11Moh44e; RGD1311922; t8130b59; TTL.6; TTLL6; Ttll6-ps1
UniProt ID: (Human) Q8N841, (Mouse) A4Q9E8
Entrez Gene ID: (Human) 284076, (Rat) 287646, (Mouse) 237930
Molecular Function:
microtubule or microtubule-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support