Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: MTTQAPTFTQ PLQSVVVLEG STATFEAHIS GFPVPEVSWF RDGQVISTST LPGVQISFSD GRAKLTIPAV TKANSGRYSL KATNGSGQAT STAELLVKAE TAPPNFVQRL
Monoclonal Anti-Titin can be used for study of the elastic filaments within sarcomeric structures. It is also useful as a differentiation marker in the separation of rhabdomyosarcomas from other muscle tumors. By indirect immunofluorescence the antibody displays a typical striated staining pattern on frozen sections of chicken skeletal and cardiac muscle tissues. Stains the region of the A-I junction by indirect immunofluorescence. It shows a decoration line 0.05 mm from the end of the A band in electron micro-graphs. In immunoblotting using total extracts of chicken breast muscle the antibody reacts specifically with both bands of the titin doublet, and showed no reaction with nebulin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2.7.11.1; Connectin; DKFZp451N061; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; Rhabdomyosarcoma antigen MU-RMS-40.14; Titin
Gene Aliases: CMD1G; CMH9; CMPD4; EOMFC; HMERF; LGMD2J; MYLK5; TMD; TTN
UniProt ID: (Human) E7EQE6
Entrez Gene ID: (Human) 7273
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support