Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELGSCLEGG REAAEEEGEP EVKKRRLLCV EFASVASCDA AVAQCFLAEN DWEMERALNS YFEPPVEESA LERRPETISE PKTYVDLTNE ETTDSTTSKI SPSEDTQQEN GSMFSLITWN IDGLDLNNLS ERARGVCSYL ALYSPDVIFL QEVIPPYYSY LKKRSSNYEI ITGHEEGYFT AIMLKKSRVK LKSQEIIPFP STKMMRNLLC VHVNVSGNEL CLMTSHLEST RGHAAERMNQ LKMVLKKMQE APESATVIFA GDTNLRDREV TRCGGLPNNI VDVWEFLGKP KHCQYTWDTQ MNSNLGITAA CKLRFDRIFF RAAAEEGHII PRSLDLLGLE KLDCGRFPSD HWGLLCNLDI IL
This gene encodes a member of a superfamily of divalent cation-dependent phosphodiesterases. The encoded protein associates with CD40, tumor necrosis factor (TNF) receptor-75 and TNF receptor associated factors (TRAFs), and inhibits nuclear factor-kappa-B activation. This protein has sequence and structural similarities with APE1 endonuclease, which is involved in both DNA repair and the activation of transcription factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-Tyr-DNA phosphodiesterase; 5'-tyrosyl-DNA phosphodiesterase; EAPII; ETS1-associated protein 2; ETS1-associated protein II; MGC111021; MGC9099; RP1-30M3.3; TRAF and TNF receptor associated protein; TRAF and TNF receptor-associated protein; Tyr-DNA phosphodiesterase 2; Tyrosyl-DNA phosphodiesterase 2; Tyrosyl-RNA phosphodiesterase; VPg unlinkase
Gene Aliases: AD-022; AD022; dJ30M3.3; EAP2; EAPII; hTDP2; TDP2; TTRAP
UniProt ID: (Human) O95551
Entrez Gene ID: (Human) 51567
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support