Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the tubulin superfamily. The encoded protein localizes to the centrosome where it binds to microtubules as part of a complex referred to as the gamma-tubulin ring complex. The protein mediates microtubule nucleation and is required for microtubule formation and progression of the cell cycle. A pseudogene of this gene is found on chromosome 7.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-1-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubulin gamma-1 chain; tubulin, gamma 1; tubulin, gamma polypeptide
Gene Aliases: CDCBM4; GCP-1; TUBG; TUBG1; TUBGCP1
UniProt ID: (Human) P23258
Entrez Gene ID: (Human) 7283
Molecular Function: tubulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support