Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gamma-ring complex protein 104 kDa; Gamma-tubulin complex component 3; GCP-3; h104p; hGCP3; hGrip104; hSpc98; spindle pole body protein; Spindle pole body protein Spc98 homolog; tubulin, gamma complex associated protein 3
Gene Aliases: 104p; GCP3; Grip104; SPBC98; Spc98; Spc98p; TUBGCP3
UniProt ID: (Human) Q96CW5, (Mouse) P58854
Entrez Gene ID: (Human) 10426, (Mouse) 259279
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support