Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: EASESTGTN SSAAHNEELS KALKGEGGTD SDHMRHEASL AIRSPCPGLE EDMEAYVLRP ALPGTMMQCY LTRDKHGVDK GLFPLYYLYL ETSDSLQRFL LAGRKRRRSK TSNYLISLDP THLSRDGDNF VGKVRSNVFS TKFTIFDNGV NPDREHLTRN TARIRQELGA VCYEPNVLGY LGPRKMTVIL PGTNSQNQRI NVQPLNEQES LLSRYQRGDK QGLLLLHNKT PSWDKENGVY TLNFHGRVTR ASVKNFQIVD PKHQEHLVLQ FGRVGPDTFT MDFCFPFSPL QAFSICLSSF N (221-520 aa encoded by BC026070)
TULP2 is a member of a family of tubby-like genes (TULPs) that encode proteins of unknown function. Members of this family have been identified in plants, vertebrates, and invertebrates. The TULP proteins share a conserved C-terminal region of approximately 200 amino acid residues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cancer testis antigen 65; Cancer/testis antigen 65; CT65; Tubby-like protein 2; Tubby-related protein 2
Gene Aliases: CT65; TUBL2; TULP2
UniProt ID: (Human) O00295
Entrez Gene ID: (Human) 7288
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support