Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tubby-like protein 3; Tubby-related protein 3
Gene Aliases: 2310022L06Rik; AI316887; TUBL3; TULP3
UniProt ID: (Human) O75386, (Mouse) O88413
Entrez Gene ID: (Human) 7289, (Mouse) 22158, (Rat) 688749
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support