Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MEASRCRLSP SGDSVFHEEM MKMRQAKLDY QRLLLEKRQR KKRLEPFMVQ PNPEARLRRA KPRASDEQTP LVNCHTPHSN VILHGIDGPA AVLKPDEVHA PSVSSSVVEE DAENTVDTAS KPGLQERLQK HDISESVNFD EETDGISQSA CLERPNSASS QNSTDTGTSG SATAAQPADN LLGDIDYLED FVYSPAPQGV TVRCRIIRDK RGMDRGLFPT YYMYLEKEEN QKIFLLAARK RKKSKTANYL ISIDPVDLSR EGESYVGKLR SNLMGTKFTV YDRGICPMKG RGLVGAAHTR QELAAISYET NVLGFKGPRK MSVIIPGMTL NHKQIPYQPQ NNHDSLLSRW QNRTMENLVE LHNKAPVWNS DTQSYVLNFR GRVTQASVKN FQIVHKNDPD YIVMQFGRVA DDVFTLDYNY PLCAVQAFGI GLSSFDSKLA CE
This gene encodes a member of the tubby-like protein (TULP) family. Members of this family have been identified in plants, vertebrates, and invertebrates, and they share a conserved C-terminal region of approximately 200 amino acid residues. The human and mouse TULP3 proteins share 69% amino acid sequence identity, with higher identity at the N and C termini than in the central region.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tubby-like protein 3; Tubby-related protein 3
Gene Aliases: TUBL3; TULP3
UniProt ID: (Human) O75386
Entrez Gene ID: (Human) 7289
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support