Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
In sandwich ELISAs, approximately 15% reactivity with recombinant mouse TWEAK is observed and no cross-reactivity with recombinant human (rh) BAFF, rhEDA-A2, rhLIGHT, rhFas Ligand, rhGITR Ligand, rhLT alpha 1 beta 2, rhLT alpha 2 beta 1, rhOX40 Ligand, rhTNF-alpha, rhTRANCE, rhTRAIL, rh4-1BB, or rhAPRIL is observed.
Reconstitute at 0.5 mg/mL in sterile PBS.
TNF-related weak inducer of apoptosis (TWEAK) is a member of the tumor necrosis factor superfamily (TNFSF) of structurally related cytokines. Like most other members of this family, TWEAK is a cell surface-associated type II transmembrane protein although a smaller, biologically active form can also be generated by cleavage near the cell membrane. TWEAK has multiple biological activities, including stimulation of cell growth and angiogenesis, induction of inflammatory cytokines, in addition to stimulation of apoptosis. The TWEAK signal transduction pathway has not been well established but it appears to signal via TweakR/Fn14 in a manner similar to that described for other TNFSF members that bind receptors lacking death domains.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: APO3 ligand; APO3/DR3 ligand; MGC129581; MGC20669; TNF-related WEAK inducer of apoptosis; tumor necrosis factor (ligand) superfamily member 12; tumor necrosis factor (ligand) superfamily, member 12; tumor necrosis factor ligand 4A; Tumor necrosis factor ligand superfamily member 12; TWEAK
Gene Aliases: APO3L; DR3LG; TNFSF12; TNLG4A; TWEAK; UNQ181/PRO207
UniProt ID: (Human) O43508
Entrez Gene ID: (Human) 8742
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support