Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Tissue lysate).
Immunogen sequence: MKLHYVAVLT LAILMFLTWL PESLSCNKAL CASDVSKCLI QELCQCRPGE GNCSCCKECM LCLGALWDEC CDCVGMCNPR NYSDTPPTSK STVEELHEPI PSLFRALTEG DTQLNWNIVS FPVAEELSHH ENLVSFLETV NQPHHQNVSV PSNNVHAPYS SDKEHMCTVV YFDDCMSIHQ CKISCESMGA SKYRWFHNAC CECIGPECID YGSKTVKCMN CMF
TWSG1 may be involved in dorsoventral axis formation. Seems to antagonize BMP signaling by forming ternary complexes with CHRD and BMPs, thereby preventing BMPs from binding to their receptors. In addition to the anti-BMP function, also has pro-BMP activity, partly mediated by cleavage and degradation of CHRD, which releases BMPs from ternary complexes. May be an important modulator of BMP-regulated cartilage development and chondrocyte differentiation. May play a role in thymocyte development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: twisted gastrulation homolog 1; Twisted gastrulation protein homolog 1
Gene Aliases: PSEC0250; TSG; TWSG1
UniProt ID: (Human) Q9GZX9
Entrez Gene ID: (Human) 57045
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support