Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Rat predicted to react based on 100% sequence homology.
Cross-reactivity in IHC with Mouse.
Thioredoxins are a group of small redox active proteins possessing a conserved active site sequence. TrxL (Thioredoxin like protein 1) has 2 distinct domains: an N terminal domain, which is 43% identical to human TXN, and a C terminal domain, which shows no homology to other proteins in the sequence databases. The active site sequence, located within the N terminal domain, is that of a thioredoxin like protein; compared to the active site sequence of TXN, it has a single amino acid substitution. Unlike other thioredoxin like proteins, TrxL does not serve as a substrate for thioredoxin reductase in an insulin assay.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 32 kDa thioredoxin-related protein; epididymis secretory protein Li 114; thioredoxin-like 1; Thioredoxin-like protein 1; thioredoxin-related 32 kDa protein; thioredoxin-related protein 1
Gene Aliases: HEL-S-114; TRP32; TXL; TXL-1; TXNL; TXNL1
UniProt ID: (Human) O43396
Entrez Gene ID: (Human) 9352
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support