Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The protein encoded by this gene contains a ubiquitin-like domain in the N-terminal region, three transmembrane segments and a C-terminal 3-oxo-5-alpha steroid 4-dehydrogenase domain. The protein belongs to the steroid 5-alpha reductase family. Mutations in this gene result in ventricular tachycardia, catecholaminergic polymorphic, 3.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: glycoprotein, synaptic 2-like; steroid 5 alpha-reductase 2-like 2; Steroid 5-alpha-reductase 2-like 2 protein; Trans-2,3-enoyl-CoA reductase-like
Gene Aliases: GPSN2L; SRD5A2L2; TECRL; TERL
UniProt ID: (Human) Q5HYJ1
Entrez Gene ID: (Human) 253017
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support