Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: VGEVENYVIV LTHFAVAGET ILVDGVSEEF RLVDLLPSTH YTATMYATNG PLTSGTISTN FSTLLDPPAN LTASEVTRQS ALISWQP
Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
Tenascin-R (TNR) is an extracellular matix protein expressed primarily in the central nervous system. It is a member of the tenascin (TN) gene family, which includes at least 3 genes in mammals: TNC (or hexabrachion; MIM 187380), TNX (TNXB; MIM 600985), and TNR (Erickson, 1993). The genes are expressed in distinct tissues at different times during embryonic development and are present in adult tissues.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Janusin; Restrictin; Tenascin-R; TN-R
Gene Aliases: TN-R; TNR
UniProt ID: (Human) Q92752
Entrez Gene ID: (Human) 7143
Molecular Function: extracellular matrix protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support