Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: GCKNYEESPE RKTLMSMPNY MQTGGLEGSH YLPPTKFSGL PRFSHDRRPS SCISWEVVEA TCACLLAQGE EAEKEHCSKC LAEQMILEEF GRCLSQILHT EFKSKGLKME
Metallothionein proteins are highly conserved low-molecular-weight cysteine-rich proteins that are induced by and bind to heavy metal ions and have no enzymatic activity. They may play a central role in the regulation of cell growth and differentiation and are involved in spermatogenesis. This gene encodes a metallothionein-like protein which has been shown to be expressed differentially in mouse testis and ovary. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CXC domain containing 2; Metallothionein-like 5, testis-specific; metallothionein-like 5, testis-specific (tesmin); Tesmin; Testis-specific metallothionein-like protein
Gene Aliases: CXCDC2; MTL5; MTLT; TESMIN
UniProt ID: (Human) Q9Y4I5
Entrez Gene ID: (Human) 9633
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support