Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%).
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
This antibody has specificity for Mouse Tetranectin/CLEC3B.
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: plasminogen kringle 4-binding protein; tetranectin; tetranectin (plasminogen binding protein); tetranectin (plasminogen-binding protein); TN
Gene Aliases: Tna
Entrez Gene ID: (Mouse) 21922
Molecular Function:
intercellular signal molecule
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support