Product References
Label-Free Quantification Mass Spectrometry Identifies Protein Markers of Chemotherapy Response in High-Grade Serous Ovarian Cancer.
Cancers
Arentz G,Mittal P,Klingler-Hoffmann M,Condina MR,Ricciardelli C,Lokman NA,Kaur G,Oehler MK,Hoffmann P
Thu Apr 06 00:00:00 EDT 2023
Thioredoxin 1 overexpression attenuated diabetes-induced endoplasmic reticulum stress in Müller cells via apoptosis signal-regulating kinase 1.
Journal of cellular biochemistry
Yu X,Teng Q,Bao K,Chudhary M,Qi H,Zhou W,Che H,Liu J,Ren X,Kong L
Wed Mar 01 00:00:00 EST 2023
Antibiotics administration alleviates the high fat diet-induced obesity through altering the lipid metabolism in young mice.
Lipids
Luo S,Zhang H,Jiang X,Xia Y,Tang S,Duan X,Sun W,Gao M,Chen C,Zou Z,Zhou L,Qiu J
Sun Jan 01 00:00:00 EST 2023
Exposure to di (2-ethylhexyl) phthalate causes locomotor increase and anxiety-like behavior via induction of oxidative stress in brain.
Toxicology mechanisms and methods
Tang S,Zhang H,Xia Y,Luo S,Liu Y,Duan X,Zou Z,Chen C,Zhou L,Qiu J
Wed Feb 01 00:00:00 EST 2023
Thioredoxin-1 regulates IRE1α to ameliorate sepsis-induced NLRP3 inflammasome activation and oxidative stress in Raw 264.7 cell.
Immunopharmacology and immunotoxicology
Shao R,Lou X,Xue J,Yang Y,Ning D,Chen G,Jiang L
Thu Jun 01 00:00:00 EDT 2023
Thioredoxin-1 regulates the autophagy induced by oxidative stress through LC3-II in human lens epithelial cells.
Clinical and experimental pharmacology & physiology
Hu J,Liu J,Chen S,Zhang C,Shen L,Yao K,Yu Y
Thu Jun 01 00:00:00 EDT 2023
Thioredoxin-1 regulates the autophagy induced by oxidative stress through LC3-II in human lens epithelial cells.
Clinical and experimental pharmacology & physiology
Hu J,Liu J,Chen S,Zhang C,Shen L,Yao K,Yu Y
Thu Jun 01 00:00:00 EDT 2023
Disruption of the lung-gut-brain axis is responsible for cortex damage induced by pulmonary exposure to zinc oxide nanoparticles.
Toxicology
Zhang L,Zhang Y,Jiang X,Mao L,Xia Y,Fan Y,Li N,Jiang Z,Qin X,Jiang Y,Liu G,Qiu F,Zhang J,Zou Z,Chen C
Wed Feb 01 00:00:00 EST 2023
Protective Effect and Mechanism of Kuntai Capsule on Angiotensin II -Induced Hypertension in Ovariectomized Rats.
Chinese journal of integrative medicine
Ge XF,Li SS,Liu YH,Lu MQ,Su HN,Yang X,Sun XW
Thu Jun 01 00:00:00 EDT 2023
Downregulation of Inflammatory Response via Nrf2/Trx1/TXNIP Axis in Oxidative Stress-Induced ARPE-19 Cells and Mouse Model of AMD.
Oxidative medicine and cellular longevity
Yang Q,Cai W,Jin H,Shen T,Yu J
Mon Dec 12 00:00:00 EST 2022
Role of thioredoxin-interacting protein in mediating endothelial dysfunction in hypertension.
Genes & diseases
Wang R,Guo Y,Li L,Luo M,Peng L,Lv D,Cheng Z,Xue Q,Wang L,Huang J
Sun May 01 00:00:00 EDT 2022
Role of thioredoxin-interacting protein in mediating endothelial dysfunction in hypertension.
Genes & diseases
Wang R,Guo Y,Li L,Luo M,Peng L,Lv D,Cheng Z,Xue Q,Wang L,Huang J
Sun May 01 00:00:00 EDT 2022
Thioredoxin-1 inhibits amyloid-β25-35-induced activation of NLRP1/caspase-1/GSDMD pyroptotic pathway in PC12 cells.
Molecular biology reports
Jia J,Zhang X,Xu G,Zeng X,Li L
Sun May 01 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Geniposide suppresses NLRP3 inflammasome-mediated pyroptosis via the AMPK signaling pathway to mitigate myocardial ischemia/reperfusion injury.
Chinese medicine
Li H,Yang DH,Zhang Y,Zheng F,Gao F,Sun J,Shi G
Fri Jun 17 00:00:00 EDT 2022
Recombinant ACE2 protein protects against acute lung injury induced by SARS-CoV-2 spike RBD protein.
Critical care (London, England)
Zhang L,Zhang Y,Qin X,Jiang X,Zhang J,Mao L,Jiang Z,Jiang Y,Liu G,Qiu J,Chen C,Qiu F,Zou Z
Thu Jun 09 00:00:00 EDT 2022
Oxidative stress as a candidate mechanism for accelerated neuroectodermal differentiation due to trisomy 21.
Free radical biology & medicine
Prutton KM,Marentette JO,Leifheit BA,Esquer H,LaBarbera DV,Anderson CC,Maclean KN,Roede JR
Wed Jun 01 00:00:00 EDT 2022
Pumpless three-dimensional photo paper-based microfluidic analytical device for automatic detection of thioredoxin-1 using enzyme-linked immunosorbent assay.
Analytical and bioanalytical chemistry
Lee MJ,Soum V,Lee SN,Choi JH,Shin JH,Shin K,Oh BK
Fri Apr 01 00:00:00 EDT 2022
LPS induces pulmonary microvascular endothelial cell barrier dysfunction by upregulating ceramide production.
Cellular signalling
Jiang J,Ouyang H,Zhou Q,Tang S,Fang P,Xie G,Yang J,Sun G
Fri Apr 01 00:00:00 EDT 2022
LPS induces pulmonary microvascular endothelial cell barrier dysfunction by upregulating ceramide production.
Cellular signalling
Jiang J,Ouyang H,Zhou Q,Tang S,Fang P,Xie G,Yang J,Sun G
Fri Apr 01 00:00:00 EDT 2022
Salidroside inhibited cerebral ischemia/reperfusion-induced oxidative stress and apoptosis via Nrf2/Trx1 signaling pathway.
Metabolic brain disease
Li F,Mao Q,Wang J,Zhang X,Lv X,Wu B,Yan T,Jia Y
Thu Dec 01 00:00:00 EST 2022
An Experimental Study Reveals the Protective Effect of Autophagy against Realgar-Induced Liver Injury via Suppressing ROS-Mediated NLRP3 Inflammasome Pathway.
International journal of molecular sciences
Yang J,Li J,Guo H,Zhang Y,Guo Z,Liu Y,Huo T
Thu May 19 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Diallyl trisulfide sensitizes radiation therapy on glioblastoma through directly targeting thioredoxin 1.
Free radical biology & medicine
Tian Y,Ge Z,Xu M,Ge X,Zhao M,Ding F,Yin J,Wang X,You Y,Shi Z,Qian X
Sat Aug 20 00:00:00 EDT 2022
Luteolin Improves Cyclophosphamide-Induced Cystitis through TXNIP/NLRP3 and NF-κB Pathways.
Evidence-based complementary and alternative medicine : eCAM
Zhang H,Zhao J,Lu Q,Sun B,Liu X,Yang C,Li S,Li L,Yi S,Yang Z,Xu J
Tue Nov 23 00:00:00 EST 2021
Lgals9 deficiency ameliorates obesity by modulating redox state of PRDX2.
Scientific reports
Nunoue T,Yamaguchi S,Teshigawara S,Katayama A,Nakatsuka A,Eguchi J,Niki T,Wada J
Tue Mar 16 00:00:00 EDT 2021
Protein S-nitrosylation regulates proteostasis and viability of hematopoietic stem cell during regeneration.
Cell reports
Yi W,Zhang Y,Liu B,Zhou Y,Liao D,Qiao X,Gao D,Xie T,Yao Q,Zhang Y,Qiu Y,Huang G,Chen Z,Chen C,Ju Z
Tue Mar 30 00:00:00 EDT 2021
Pulmonary Exposure to Copper Oxide Nanoparticles Leads to Neurotoxicity via Oxidative Damage and Mitochondrial Dysfunction.
Neurotoxicity research
Zhou H,Yao L,Jiang X,Sumayyah G,Tu B,Cheng S,Qin X,Zhang J,Zou Z,Chen C
Sun Aug 01 00:00:00 EDT 2021
Sub-chronic exposure to realgar induces liver injury via upregulating the TXNIP/NLRP3 pathway and disturbing bile acid homeostasis in mice.
Journal of ethnopharmacology
Li A,Wu X,Yang J,Li J,Guo H,Zhang Y,Jiang H,Huo T
Sun Dec 05 00:00:00 EST 2021
Verapamil ameliorates proximal tubular epithelial cells apoptosis and fibrosis in diabetic kidney.
European journal of pharmacology
Song Y,Guo F,Zhao Y,Zhao L,Fan X,Zhang Y,Liu Y,Qin G
Mon Nov 15 00:00:00 EST 2021
Thioredoxin-1 Rescues MPP+/MPTP-Induced Ferroptosis by Increasing Glutathione Peroxidase 4.
Molecular neurobiology
Bai L,Yan F,Deng R,Gu R,Zhang X,Bai J
Thu Jul 01 00:00:00 EDT 2021
Stabilization of Nrf2 leading to HO-1 activation protects against zinc oxide nanoparticles-induced endothelial cell death.
Nanotoxicology
Zhang L,Zou L,Jiang X,Cheng S,Zhang J,Qin X,Qin Z,Chen C,Zou Z
Sun Aug 01 00:00:00 EDT 2021
Upregulation of thioredoxin contributes to inhibiting diabetic hearing impairment.
Diabetes research and clinical practice
Ren X,Lv J,Fu Y,Zhang N,Zhang C,Dong Z,Chudhary M,Zhong S,Kong L,Kong H
Wed Sep 01 00:00:00 EDT 2021
Libertellenone H, a Natural Pimarane Diterpenoid, Inhibits Thioredoxin System and Induces ROS-Mediated Apoptosis in Human Pancreatic Cancer Cells.
Molecules (Basel, Switzerland)
Zhang W,Zhu Y,Yu H,Liu X,Jiao B,Lu X
Sat Jan 09 00:00:00 EST 2021
Methods to separate nuclear soluble fractions reflecting localizations in living cells.
iScience
Ogawa Y,Imamoto N
Fri Dec 17 00:00:00 EST 2021
Methods to separate nuclear soluble fractions reflecting localizations in living cells.
iScience
Ogawa Y,Imamoto N
Fri Dec 17 00:00:00 EST 2021
Preservation of mitochondrial homeostasis is responsible for the ameliorative effects of Suhuang antitussive capsule on non-resolving inflammation via inhibition of NF-κB signaling and NLRP3 inflammasome activation.
Journal of ethnopharmacology
Qin W,Tong X,Liang R,Tang K,Wu X,Jia Y,Tan N
Mon May 10 00:00:00 EDT 2021
Preservation of mitochondrial homeostasis is responsible for the ameliorative effects of Suhuang antitussive capsule on non-resolving inflammation via inhibition of NF-κB signaling and NLRP3 inflammasome activation.
Journal of ethnopharmacology
Qin W,Tong X,Liang R,Tang K,Wu X,Jia Y,Tan N
Mon May 10 00:00:00 EDT 2021
TFE3, a potential therapeutic target for Spinal Cord Injury via augmenting autophagy flux and alleviating ER stress.
Theranostics
Zhou K,Zheng Z,Li Y,Han W,Zhang J,Mao Y,Chen H,Zhang W,Liu M,Xie L,Zhang H,Xu H,Xiao J
Fri May 28 00:00:00 EDT 2021
WJ-39, an Aldose Reductase Inhibitor, Ameliorates Renal Lesions in Diabetic Nephropathy by Activating Nrf2 Signaling.
Oxidative medicine and cellular longevity
Zhou X,Liu Z,Ying K,Wang H,Liu P,Ji X,Chi T,Zou L,Wang S,He Z
Wed Jan 27 00:00:00 EST 2021
Sulforaphane attenuates apoptosis of hippocampal neurons induced by high glucose via regulating endoplasmic reticulum.
Neurochemistry international
Tang L,Ren X,Han Y,Chen L,Meng X,Zhang C,Chu H,Kong L,Ma H
Mon Jun 01 00:00:00 EDT 2020
Targeting the MAPK/ERK and PI3K/AKT Signaling Pathways Affects NRF2, Trx and GSH Antioxidant Systems in Leukemia Cells.
Antioxidants (Basel, Switzerland)
Jasek-Gajda E,Jurkowska H,Jasińska M,Lis GJ
Fri Jul 17 00:00:00 EDT 2020
Neuroprotective effects of notoginsenoside R1 by upregulating Trx-1 on acrylamide-induced neurotoxicity in PC12.
Human & experimental toxicology
Wang W,Huang L,Hu Y,Thomas ER,Li X
Mon Jun 01 00:00:00 EDT 2020
Novel Oxindole-Curcumin Hybrid Compound for Antioxidative Stress and Neuroprotection.
ACS chemical neuroscience
Hirata Y,Ito Y,Takashima M,Yagyu K,Oh-Hashi K,Suzuki H,Ono K,Furuta K,Sawada M
Thu Jan 02 00:00:00 EST 2020
Icariin Prevents Extracellular Matrix Accumulation and Ameliorates Experimental Diabetic Kidney Disease by Inhibiting Oxidative Stress via GPER Mediated p62-Dependent Keap1 Degradation and Nrf2 Activation.
Frontiers in cell and developmental biology
Wang K,Zheng X,Pan Z,Yao W,Gao X,Wang X,Ding X
Mon Sep 28 00:00:00 EDT 2020
Notoginsenoside R1 Protects Against the Acrylamide-Induced Neurotoxicity via Upregulating Trx-1-Mediated ITGAV Expression: Involvement of Autophagy.
Frontiers in pharmacology
Wang W,Huang L,Thomas ER,Hu Y,Zeng F,Li X
Thu Oct 01 00:00:00 EDT 2020
Triptolide impairs thioredoxin system by suppressing Notch1-mediated PTEN/Akt/Txnip signaling in hepatocytes.
Toxicology letters
Shen F,Xiong Z,Kong J,Wang L,Cheng Y,Jin J,Huang Z
Tue Jan 01 00:00:00 EST 2019
Triptolide impairs thioredoxin system by suppressing Notch1-mediated PTEN/Akt/Txnip signaling in hepatocytes.
Toxicology letters
Shen F,Xiong Z,Kong J,Wang L,Cheng Y,Jin J,Huang Z
Tue Jan 01 00:00:00 EST 2019
Cordycepin prevents radiation ulcer by inhibiting cell senescence via NRF2 and AMPK in rodents.
Nature communications
Wang Z,Chen Z,Jiang Z,Luo P,Liu L,Huang Y,Wang H,Wang Y,Long L,Tan X,Liu D,Jin T,Wang Y,Wang Y,Liao F,Zhang C,Chen L,Gan Y,Liu Y,Yang F,Huang C,Miao H,Chen J,Cheng T,Fu X,Shi C
Mon Jun 10 00:00:00 EDT 2019
Detection of thioredoxin-1 using ultra-sensitive ELISA with enzyme-encapsulated human serum albumin nanoparticle.
Nano convergence
Lee MJ,Lee ES,Kim TH,Jeon JW,Kim Y,Oh BK
Mon Dec 09 00:00:00 EST 2019
FOXO1 Overexpression Attenuates Tubulointerstitial Fibrosis and Apoptosis in Diabetic Kidneys by Ameliorating Oxidative Injury via TXNIP-TRX.
Oxidative medicine and cellular longevity
Ji L,Wang Q,Huang F,An T,Guo F,Zhao Y,Liu Y,He Y,Song Y,Qin G
Tue Jul 09 00:00:00 EDT 2019
FOXO1 Overexpression Attenuates Tubulointerstitial Fibrosis and Apoptosis in Diabetic Kidneys by Ameliorating Oxidative Injury via TXNIP-TRX.
Oxidative medicine and cellular longevity
Ji L,Wang Q,Huang F,An T,Guo F,Zhao Y,Liu Y,He Y,Song Y,Qin G
Tue Jul 09 00:00:00 EDT 2019
FOXO1 Overexpression Attenuates Tubulointerstitial Fibrosis and Apoptosis in Diabetic Kidneys by Ameliorating Oxidative Injury via TXNIP-TRX.
Oxidative medicine and cellular longevity
Ji L,Wang Q,Huang F,An T,Guo F,Zhao Y,Liu Y,He Y,Song Y,Qin G
Tue Jul 09 00:00:00 EDT 2019
FOXO1 Overexpression Attenuates Tubulointerstitial Fibrosis and Apoptosis in Diabetic Kidneys by Ameliorating Oxidative Injury via TXNIP-TRX.
Oxidative medicine and cellular longevity
Ji L,Wang Q,Huang F,An T,Guo F,Zhao Y,Liu Y,He Y,Song Y,Qin G
Tue Jul 09 00:00:00 EDT 2019
Neuroprotective effects of Brazilian green propolis on oxytosis/ferroptosis in mouse hippocampal HT22 cells.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association
Takashima M,Ichihara K,Hirata Y
Tue Oct 01 00:00:00 EDT 2019
Cardamonin, a natural flavone, alleviates inflammatory bowel disease by the inhibition of NLRP3 inflammasome activation via an AhR/Nrf2/NQO1 pathway.
Biochemical pharmacology
Wang K,Lv Q,Miao YM,Qiao SM,Dai Y,Wei ZF
Sat Sep 01 00:00:00 EDT 2018
Cardamonin, a natural flavone, alleviates inflammatory bowel disease by the inhibition of NLRP3 inflammasome activation via an AhR/Nrf2/NQO1 pathway.
Biochemical pharmacology
Wang K,Lv Q,Miao YM,Qiao SM,Dai Y,Wei ZF
Sat Sep 01 00:00:00 EDT 2018
Hypothermic oxygenated perfusion (HOPE) attenuates ischemia/reperfusion injury in the liver through inhibition of the TXNIP/NLRP3 inflammasome pathway in a rat model of donation after cardiac death.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
He W,Ye S,Zeng C,Xue S,Hu X,Zhang X,Gao S,Xiong Y,He X,Vivalda S,Li L,Wang Y,Ye Q
Tue Jun 05 00:00:00 EDT 2018
Hypothermic oxygenated perfusion (HOPE) attenuates ischemia/reperfusion injury in the liver through inhibition of the TXNIP/NLRP3 inflammasome pathway in a rat model of donation after cardiac death.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology
He W,Ye S,Zeng C,Xue S,Hu X,Zhang X,Gao S,Xiong Y,He X,Vivalda S,Li L,Wang Y,Ye Q
Tue Jun 05 00:00:00 EDT 2018
Geranylgeranylacetone blocks the reinstatement of morphine-conditioned place preference.
Neuropharmacology
Guo N,Zhang X,Huang M,Li X,Li Y,Zhou X,Bai J
Sat Dec 01 00:00:00 EST 2018
Geranylgeranylacetone blocks the reinstatement of morphine-conditioned place preference.
Neuropharmacology
Guo N,Zhang X,Huang M,Li X,Li Y,Zhou X,Bai J
Sat Dec 01 00:00:00 EST 2018
Overexpression of Thioredoxin-1 Blocks Morphine-Induced Conditioned Place Preference Through Regulating the Interaction of γ-Aminobutyric Acid and Dopamine Systems.
Frontiers in neurology
Li X,Huang M,Yang L,Guo N,Yang X,Zhang Z,Bai M,Ge L,Zhou X,Li Y,Bai J
Mon Mar 21 00:00:00 EDT 2022
Overexpression of Thioredoxin-1 Blocks Morphine-Induced Conditioned Place Preference Through Regulating the Interaction of γ-Aminobutyric Acid and Dopamine Systems.
Frontiers in neurology
Li X,Huang M,Yang L,Guo N,Yang X,Zhang Z,Bai M,Ge L,Zhou X,Li Y,Bai J
Mon Mar 21 00:00:00 EDT 2022
Thioredoxin-1 Protects Spinal Cord from Demyelination Induced by Methamphetamine through Suppressing Endoplasmic Reticulum Stress and Inflammation.
Frontiers in neurology
Yang L,Guo Y,Huang M,Wu X,Li X,Chen G,Li Y,Bai J
Wed Sep 30 00:00:00 EDT 2020
Thioredoxin-1 Protects Spinal Cord from Demyelination Induced by Methamphetamine through Suppressing Endoplasmic Reticulum Stress and Inflammation.
Frontiers in neurology
Yang L,Guo Y,Huang M,Wu X,Li X,Chen G,Li Y,Bai J
Wed Sep 30 00:00:00 EDT 2020
Mild Hypothermia Protects Renal Function in Ischemia-reperfusion Kidney: An Experimental Study in Mice.
Transplantation proceedings
Xia Z,Wang W,Xiao Q,Ye Q,Zhang X,Wang Y
Sat Dec 01 00:00:00 EST 2018
Aberrant expression of redox regulatory proteins in patients with concomitant primary Sclerosing cholangitis/inflammatory bowel disease.
Experimental and molecular pathology
Petersen DR,Orlicky DJ,Roede JR,Shearn CT
Wed Aug 01 00:00:00 EDT 2018
Aberrant expression of redox regulatory proteins in patients with concomitant primary Sclerosing cholangitis/inflammatory bowel disease.
Experimental and molecular pathology
Petersen DR,Orlicky DJ,Roede JR,Shearn CT
Wed Aug 01 00:00:00 EDT 2018
Thioredoxin silencing-induced cardiac supercontraction occurs through endoplasmic reticulum stress and calcium overload in chicken.
Metallomics : integrated biometal science
Yang J,Gong Y,Liu Q,Cai J,Zhang B,Zhang Z
Wed Nov 14 00:00:00 EST 2018
Differential carbonylation of proteins in end-stage human fatty and nonfatty NASH.
Free radical biology & medicine
Shearn CT,Saba LM,Roede JR,Orlicky DJ,Shearn AH,Petersen DR
Fri Dec 01 00:00:00 EST 2017
Differential carbonylation of proteins in end-stage human fatty and nonfatty NASH.
Free radical biology & medicine
Shearn CT,Saba LM,Roede JR,Orlicky DJ,Shearn AH,Petersen DR
Fri Dec 01 00:00:00 EST 2017
Differential carbonylation of proteins in end-stage human fatty and nonfatty NASH.
Free radical biology & medicine
Shearn CT,Saba LM,Roede JR,Orlicky DJ,Shearn AH,Petersen DR
Fri Dec 01 00:00:00 EST 2017
Differential carbonylation of proteins in end-stage human fatty and nonfatty NASH.
Free radical biology & medicine
Shearn CT,Saba LM,Roede JR,Orlicky DJ,Shearn AH,Petersen DR
Fri Dec 01 00:00:00 EST 2017
Selenium deficiency-induced thioredoxin suppression and thioredoxin knock down disbalanced insulin responsiveness in chicken cardiomyocytes through PI3K/Akt pathway inhibition.
Cellular signalling
Yang J,Hamid S,Cai J,Liu Q,Xu S,Zhang Z
Sun Oct 01 00:00:00 EDT 2017
Selenium deficiency-induced thioredoxin suppression and thioredoxin knock down disbalanced insulin responsiveness in chicken cardiomyocytes through PI3K/Akt pathway inhibition.
Cellular signalling
Yang J,Hamid S,Cai J,Liu Q,Xu S,Zhang Z
Sun Oct 01 00:00:00 EDT 2017
SIRT1 Regulates the Chemoresistance and Invasiveness of Ovarian Carcinoma Cells.
Translational oncology
Mvunta DH,Miyamoto T,Asaka R,Yamada Y,Ando H,Higuchi S,Ida K,Kashima H,Shiozawa T
Tue Aug 01 00:00:00 EDT 2017
SIRT1 Regulates the Chemoresistance and Invasiveness of Ovarian Carcinoma Cells.
Translational oncology
Mvunta DH,Miyamoto T,Asaka R,Yamada Y,Ando H,Higuchi S,Ida K,Kashima H,Shiozawa T
Tue Aug 01 00:00:00 EDT 2017
Gene expression of selenoproteins can be regulated by thioredoxin(Txn) silence in chicken cardiomyocytes.
Journal of inorganic biochemistry
Yang J,Hamid S,Liu Q,Cai J,Xu S,Zhang Z
Fri Dec 01 00:00:00 EST 2017
Gene expression of selenoproteins can be regulated by thioredoxin(Txn) silence in chicken cardiomyocytes.
Journal of inorganic biochemistry
Yang J,Hamid S,Liu Q,Cai J,Xu S,Zhang Z
Fri Dec 01 00:00:00 EST 2017
Effects of Hypobaric Hypoxia on Rat Retina and Protective Response of Resveratrol to the Stress.
International journal of medical sciences
Xin X,Dang H,Zhao X,Wang H
Mon May 21 00:00:00 EDT 2018
Effects of Hypobaric Hypoxia on Rat Retina and Protective Response of Resveratrol to the Stress.
International journal of medical sciences
Xin X,Dang H,Zhao X,Wang H
Mon May 21 00:00:00 EDT 2018
Glutaredoxins concomitant with optimal ROS activate AMPK through S-glutathionylation to improve glucose metabolism in type 2 diabetes.
Free radical biology & medicine
Dong K,Wu M,Liu X,Huang Y,Zhang D,Wang Y,Yan LJ,Shi D
Thu Dec 01 00:00:00 EST 2016
Glutaredoxins concomitant with optimal ROS activate AMPK through S-glutathionylation to improve glucose metabolism in type 2 diabetes.
Free radical biology & medicine
Dong K,Wu M,Liu X,Huang Y,Zhang D,Wang Y,Yan LJ,Shi D
Thu Dec 01 00:00:00 EST 2016
Tubeimoside-1 induces oxidative stress-mediated apoptosis and G0/G1 phase arrest in human prostate carcinoma cells in vitro.
Acta pharmacologica Sinica
Yang JB,Khan M,He YY,Yao M,Li YM,Gao HW,Ma TH
Fri Jul 01 00:00:00 EDT 2016
Tubeimoside-1 induces oxidative stress-mediated apoptosis and G0/G1 phase arrest in human prostate carcinoma cells in vitro.
Acta pharmacologica Sinica
Yang JB,Khan M,He YY,Yao M,Li YM,Gao HW,Ma TH
Fri Jul 01 00:00:00 EDT 2016
Ischemic postconditioning relieves cerebral ischemia and reperfusion injury through activating T-LAK cell-originated protein kinase/protein kinase B pathway in rats.
Stroke
Zhao H,Wang R,Tao Z,Gao L,Yan F,Gao Z,Liu X,Ji X,Luo Y
Fri Aug 01 00:00:00 EDT 2014
Ischemic postconditioning relieves cerebral ischemia and reperfusion injury through activating T-LAK cell-originated protein kinase/protein kinase B pathway in rats.
Stroke
Zhao H,Wang R,Tao Z,Gao L,Yan F,Gao Z,Liu X,Ji X,Luo Y
Fri Aug 01 00:00:00 EDT 2014
Altered dietary methionine differentially impacts glutathione and methionine metabolism in long-living growth hormone-deficient Ames dwarf and wild-type mice.
Longevity & healthspan
Brown-Borg HM,Rakoczy S,Wonderlich JA,Armstrong V,Rojanathammanee L
Tue Jan 13 00:00:00 EST 2015
Altered dietary methionine differentially impacts glutathione and methionine metabolism in long-living growth hormone-deficient Ames dwarf and wild-type mice.
Longevity & healthspan
Brown-Borg HM,Rakoczy S,Wonderlich JA,Armstrong V,Rojanathammanee L
Tue Jan 13 00:00:00 EST 2015
The Ets transcription factor GABP is a component of the hippo pathway essential for growth and antioxidant defense.
Cell reports
Wu H,Xiao Y,Zhang S,Ji S,Wei L,Fan F,Geng J,Tian J,Sun X,Qin F,Jin C,Lin J,Yin ZY,Zhang T,Luo L,Li Y,Song S,Lin SC,Deng X,Camargo F,Avruch J,Chen L,Zhou D
Thu May 30 00:00:00 EDT 2013
The Ets transcription factor GABP is a component of the hippo pathway essential for growth and antioxidant defense.
Cell reports
Wu H,Xiao Y,Zhang S,Ji S,Wei L,Fan F,Geng J,Tian J,Sun X,Qin F,Jin C,Lin J,Yin ZY,Zhang T,Luo L,Li Y,Song S,Lin SC,Deng X,Camargo F,Avruch J,Chen L,Zhou D
Thu May 30 00:00:00 EDT 2013
Interaction between M-like protein and macrophage thioredoxin facilitates antiphagocytosis for Streptococcus equi ssp. zooepidemicus.
PloS one
Ma Z,Zhang H,Zheng J,Li Y,Yi L,Fan H,Lu C
Mon Aug 27 00:00:00 EDT 2012
Interaction between M-like protein and macrophage thioredoxin facilitates antiphagocytosis for Streptococcus equi ssp. zooepidemicus.
PloS one
Ma Z,Zhang H,Zheng J,Li Y,Yi L,Fan H,Lu C
Mon Aug 27 00:00:00 EDT 2012
Interaction between M-like protein and macrophage thioredoxin facilitates antiphagocytosis for Streptococcus equi ssp. zooepidemicus.
PloS one
Ma Z,Zhang H,Zheng J,Li Y,Yi L,Fan H,Lu C
Mon Aug 27 00:00:00 EDT 2012
Interaction between M-like protein and macrophage thioredoxin facilitates antiphagocytosis for Streptococcus equi ssp. zooepidemicus.
PloS one
Ma Z,Zhang H,Zheng J,Li Y,Yi L,Fan H,Lu C
Mon Aug 27 00:00:00 EDT 2012
Overexpression of thioredoxin system proteins predicts poor prognosis in patients with squamous cell carcinoma of the tongue.
Oral oncology
Zhu X,Huang C,Peng B
Fri Jul 01 00:00:00 EDT 2011
Overexpression of thioredoxin system proteins predicts poor prognosis in patients with squamous cell carcinoma of the tongue.
Oral oncology
Zhu X,Huang C,Peng B
Fri Jul 01 00:00:00 EDT 2011