Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
The target is usually found in the following locations: Secreted.
Immunogen sequence: STLSFYQRRR FSPDDSAGAS ALLRSGPYMP QCDAFGSWEP VQCHAGTGHC WCVDEKGGFI PGSLTARSLQ IPQCPTTCEK SRTSGLLSSW KQARSQENPS P
The human thyroglobulin (hTg) is a high molecular weight glycoprotein (605 kDa) found in the thyroid follicular cells. It plays a central role in the uptake, incorporation, and regulated biosynthesis of thyroid hormones, T4 and T3. Thyroid disorders are, in large part, due to autoimmune origin, and anti-thyroglobulin autoantibodies were the first factor to be discovered. Anti-hTG is found in all thyroid autoimmune diseases (Hashimoto thyroiditis, Graves diseases), with the highest level observed in Hashimoto thyroiditis. Anti-hTG is also characteristic of thyroid cancer, and its determination can be used for the follow up of cancer patients.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Colloid; congenital goiter; Tg; Thyroglobulin
Gene Aliases: AITD3; cog; TG; TGN
UniProt ID: (Human) P01266, (Mouse) O08710
Entrez Gene ID: (Human) 7038, (Mouse) 21819, (Rat) 24826
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support