Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRMAQAFQN VREEPAVQFN SGTLALNRKV KNNPDPTIYP VLDWNDIKFQ DVIGEGNFGQ VLKARIKKDG LRMDAAIKRM KEYASKDDHR DFAGELEVLC KLGHHPNIIN LLGACEHRGY LYLAIEYAPH GNLLDFLRKS RVLETDPAFA IANSTASTLS SHHLLHFAAD VARGMDYLSQ KQFIHRDLAA RNILVGENYV AKIADFGLSR GQEVYVKKTM GRLPVRWMAI ESLNYSVYTT NSDVWSYGVL LWEIVSLGGT PYCGMTCAEL YEKLPQGYRL EKPLNCDDEV YDLMRQCWRE KPYERPSFAQ ILVSLNRMLE ERKTYVNTTL YEKFTYAGID CSAEEAA (779-1124 aa encoded by BC035514)
The TEK receptor tyrosine kinase is expressed almost exclusively in endothelial cells. TEK signaling appears to be critical for endothelial cell-smooth muscle cell communication in venous morphogenesis, and defects in TEK are associated with inherited venous malformations. The ligand angiopoietin-1 binds to this receptor and mediates a signaling pathway that functions in embryonic vascular development. Immunoblotting showed that TIE2 expression was increased by thyroid-stimulating hormone and agents that increased intracellular cAMP. HSCs expressing the receptor tyrosine kinase TIE2 are quiescent and antiapoptotic and comprise a side population of HSCs that adhere to osteoblasts in the bone marrow niche.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Angiopoietin-1 receptor; CD202b; CD202b antigen; EC 2.7.10; EC 2.7.10.1; Endothelial tyrosine kinase; endothelial-specific receptor tyrosine kinase; hTIE2; HYK; mTIE2; p140 TEK; STK1; TEK tyrosine kinase, endothelial; TEKT1; Tunica interna endothelial cell kinase; Tyrosine kinase with Ig and EGF homology domains-2; tyrosine kinases that contain immunoglobulin-like loops and epidermal growth factor-similar domains 2; Tyrosine-protein kinase receptor TEK; Tyrosine-protein kinase receptor TIE-2
Gene Aliases: AA517024; CD202B; Hyk; STK1; TEK; TIE-2; TIE2; VMCM; VMCM1
UniProt ID: (Human) Q8IV34, (Mouse) Q02858
Entrez Gene ID: (Human) 7010, (Mouse) 21687, (Rat) 89804
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support