Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Transketolase encodes a thiamine-dependent enzyme which plays a role in the channeling of excess sugar phosphates to glycolysis in the pentose phosphate pathway. Multiple alternatively spliced variants, encoding the same protein, have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 107; epididymis secretory protein Li 48; TK; Transketolase
Gene Aliases: HEL-S-48; HEL107; TK; TKT; TKT1
UniProt ID: (Human) P29401
Entrez Gene ID: (Human) 7086
Molecular Function:
transketolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support