Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Specificity: Specific for T. vaginalis.
Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 µg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: t vaginalis; T. vaginalis
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support