Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TIF1 gamma is a member of the Transcriptional Intermediary Factor 1 family, which serve as coactivators and corepressors. Its structure is similar to TIF1 alpha and TIF1 beta, exhibiting multiple domains (RING finger, coiled coil, B boxes, PHD/TTC, and bromodomain). TIF1 gamma functions in cell differentiation and development, playing a role in differentiation of hematopoietic cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM33; Ectodermin homolog; Protein Rfg7; RET-fused gene 7 protein; RING-type E3 ubiquitin transferase TRIM33; TIF 1-g; TIF1-gamma; Transcription intermediary factor 1-gamma; transcriptional intermediary factor 1 gamma; tripartite motif-containing 33; Tripartite motif-containing protein 33
Gene Aliases: ECTO; KIAA1113; PTC7; RFG7; TF1G; TIF1G; TIF1GAMMA; TIFGAMMA; TRIM33
UniProt ID: (Human) Q9UPN9
Entrez Gene ID: (Human) 51592
Molecular Function: chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support