Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: CVCSKTEIN CRRPDDGNLF PLLEGQDSGN SNGNASINIT DISRNITSIH IENWRSLHTL NAVDMELYTG LQKLTIKNSG LRSIQPRAFA KNPHLRYINL SSNRLTTLSW QLFQTLSLRE LQLEQNFFNC SCDIRWMQLW QEQGEAKLNS QNLYCINADG SQLPLFRMNI SQCDLPEISV SHVNLTVREG DNAVITCNGS GSPLPDVDWI VTGLQSINTH QTNLNWTNVH AINLTLVNVT SEDNGFTLTC IAENVVGMSN ASVALTVYYP PRVVSLEEPE LRLEHCIEFV VRGNPPPTLH WLHNGQPLRE SKIIHV (36-350 aa encoded by BC013693)
NTRK3 (TRKC) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. Known substrates for the TRK receptors are SHC, PI-3 kinase, and PLCG1. The different isoforms do not have identical signaling properties. The protein is widely expressed, mainly in the nervous tissue. The isoform B is expressed in a relatively large amount in the adult brain comparatively to fetal brain. TRKC is subject to ligand-mediated auto-phosphorylation. The protein structure contains 2 immunoglobulin-like C2-type domains and 2 leucine-rich (LRR) repeats.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; GP145-TrkC; neural receptor protein-tyrosine kinase (trkC); Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 3; neurotrophin-3; NT-3 growth factor receptor; Trk-C; TrkC tyrosine kinase; tyrosine kinase receptor; tyrosine kinase receptor C
Gene Aliases: AW125844; gp145(trkC); GP145-TrkC; NTRK3; Ntrk3_tv3; TRKC
UniProt ID: (Pig) P24786, (Human) Q16288, (Mouse) Q6VNS1, (Rat) Q03351
Entrez Gene ID: (Pig) 397511, (Human) 4916, (Mouse) 18213, (Rat) 29613
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support